Learn More
Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein
Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water or appropriate buffered solution containing 0.1% BSA to regain full activity. Apportion there constituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in buffered solution containing a carrier protein, such as PBS + 0.1% BSA. Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Accession Number | P49771 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 2323 |
| Molecular Weight (g/mol) | 35 kDa |
| Name | Human Flt-3 Ligand (FLT3L) |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.