missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Flt-3 Ligand (FLT3L), Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Human Flt3-Ligand is a soluble 17.6 kDa protein consisting of 155 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
Specifications
Specifications
| Accession Number | P49771 |
| For Use With (Application) | Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 2323 |
| Molecular Weight (g/mol) | 17.6 kDa |
| Name | Human Flt-3 Ligand (FLT3L), Animal-Free |
| Purification Method | Purified |
| Quantity | 2 μg |
| Storage Requirements | -20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?