missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Flt-3/Flk-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£161.00 - £406.00
Specifications
| Antigen | Flt-3/Flk-2/CD135 |
|---|---|
| Clone | 1038631 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704687
![]() |
R&D Systems
MAB8122-SP |
25 μg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790707
![]() |
R&D Systems
MAB8122-100 |
100 μg |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for ImmunohistochemistrySpécification
| Flt-3/Flk-2/CD135 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, Flk-2, FLK2, Flt3, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| Human embryonic kidney cell HEK293-derived human Flt-3/Flk-2, Met1-Asn541, Accession # P36888 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1038631 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P36888 | |
| 2322 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit