missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FLRT3 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Dimensione della confezione:
Each
Pack of 96
Descrizione
Human FLRT3 quantitates human FLRT3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FLRT3.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRTs may function in cell adhesion and/or receptor signaling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. This gene is expressed in many tissues. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
Specifica
Specifica
| Accession Number | 5530600M07Rik; C430047I10Rik; Fibronectin leucine rich transmembrane protein 3; Fibronectin-like domain-containing leucine-rich transmembrane protein 3; FLRT3; HH21; KIAA1469; Leucine-rich repeat transmembrane protein FLRT3; mKIAA1469; RGD1566005; UNQ856/PRO1865 |
| Assay Range | 0.14 to 25 ng/mL |
| Assay Sensitivity | 0.14 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 23767 |
| Gene Symbol | FLRT3 |
| Interassay CV | <12% |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto