missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FLRT1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2794
Additional Details : Weight : 0.00970kg
Description
FLRT1 Polyclonal specifically detects FLRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
FLRT1 | |
Polyclonal | |
Unconjugated | |
Q9NZU1 | |
FLRT1 | |
Mouse myeloma cell line NS0-derived recombinant human FLRT1 Ile21-Pro524 Accession # Q9NZU1 | |
100 μg | |
Primary | |
Detects human FLRT1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) FLRT3 is observed and less than 1% cross-reactivity with rhFLRT2 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624, SPG68 | |
Goat | |
Affinity Purified | |
RUO | |
23769 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only