missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Recognizes human FKBPL
- FKBPL (DIR1, NG7, WISp39) similar to immunophilin protein family, which plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking
- Thought to have potential role in induced radioresistance
- Breast cancer cells stably overexpressing FKBPL became dependent on estrogen for growth
- FKBPL is an estrogen-inducible gene that acts as a co-chaperone in ERa/Hsp90 molecular complexes
- Levels may be prognostic indicator and determinant of response to endocrine therapy
Process:
- Antibody specific for FKBPL pre-coated onto microwells
- FKBPL protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody specific for FKBPL is added to detect captured FKBPL protein
- For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, meaurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | NP_071393 |
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 7 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 63943 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human APCS ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?