missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FKBP12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB37771-100
This item is not returnable.
View return policy
Description
FKBP12 Monoclonal antibody specifically detects FKBP12 in Human samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| FKBP12 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP1a, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PKC12, PPIASE, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| E. coli-derived human FKBP12F36V protein, Gly1-Glu108, Accession # P62942 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 1049713 | |
| Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| P62942 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2280 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction