missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FKBP10 (aa 92-184) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144711 (PA5-144711. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.
Specifications
Specifications
| Accession Number | Q96AY3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 60681 |
| Name | Human FKBP10 (aa 92-184) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 65 kDa FK506-binding protein; 65 kDa FKBP; AI325255; BRKS1; FK506 binding protein 10; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP10; FKBP-10; Fkbp1-rs; Fkbp6; FKBP65; FKBP-65; Fkbprp; Fkbp-rs; Fkbp-rs1; hFKBP65; Immunophilin FKBP65; OI11; OI6; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIASE; PPIase FKBP10; PSEC0056; Rotamase |
| Common Name | FKBP10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction