missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ficolin-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2367
This item is not returnable.
View return policy
Description
Ficolin-3 Isoform 2 Polyclonal specifically detects Ficolin-3 Isoform 2 in Human samples. It is validated for Western Blot.
Specifications
| Ficolin-3 Isoform 2 | |
| Polyclonal | |
| Unconjugated | |
| O75636-1 | |
| FCN3 | |
| Mouse myeloma cell line NS0-derived recombinant human Ficolin-3 Lys22-Arg299 Accession # O75636-1 | |
| 100 μg | |
| Primary | |
| Detects human Ficolin-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) Ficolin-1 is observed and less 1% cross-reactivity with rhFicolin-2 and rhTenascinR is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| FCN3, FCNH, Ficolin3, HAKA1, H-Ficolin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8547 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction