missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fibronectin Coated 96-well Microplates, 5 Pack

Product Code. 18471150
Change view
Click to view available options
Quantity:
5/Pk.
Unit Size:
1 set
Product Code. Quantity unitSize
18471150 5/Pk. 1 set
1 options
This item is not returnable. View return policy

Product Code. 18471150

Brand: R&D Systems™ CWP001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides solid support for cell attachment of adherent cell lines using serum-derived and filtered human fibronectin

Fibronectin is an extracellular matrix (ECM) component and is one of the primary cell adhesion molecules. The occurrence of different isoforms is due to alternative mRNA splicing of the ED-A, ED-B and III-CS regions, and subsequent post-translational modification. Although non-reactive with adhesion receptors in its soluble state, fibronectin is highly adhesive when immobilized on a surface.

Clear, polystyrene tissue culture microplates with 96 wells
Coated with 0.2 micron filtered serum-derived human fibronectin (FN) and/or blocked with Bovine Serum Albumin (BSA) Fraction V under aseptic conditions
Derived from human plasma nonreactive for HIV-1/2 antibodies and HbsAg at the donor level

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Array 8 x 12
No. of Wells 96
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.