missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibroblast Activation Protein alpha/FAP APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3715A-100
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry.
Specifications
| Fibroblast Activation Protein alpha/FAP | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 427819 | |
| Flow Cytometry 10 μL/106 cells | |
| Q12884 | |
| FAP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP, Leu26-Asp760, Accession # Q12884 | |
| 100 Tests | |
| 2191 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction