missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™
Description
Fibroblast Activation Protein alpha/FAP Polyclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.
Specifications
Specifications
| Antigen | Fibroblast Activation Protein alpha/FAP |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoassay |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated 0.5 ug/mL |
| Gene Accession No. | Q12884 |
| Gene Alias | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
| Gene Symbols | FAP |
| Host Species | Sheep |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?