missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FHL1 (aa 34-135) Control Fragment Recombinant Protein

Product Code. 30209854
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209854

Brand: Invitrogen™ RP93667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51503 (PA5-51503. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13642
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2273
Name Human FHL1 (aa 34-135) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1 A; four and a half LIM domains 1 b; four and a half LIM domains 1 b; four and a half LIM domains protein 1 b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1 A; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBM x 1 A; RBM x 1 B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025
Common Name FHL1
Gene Symbol FHL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KFCANTCVECRKPIGADSKEVHYKNRFWHDTCFRCAKCLHPLANETFVAKDNKILCNKCTTREDSPKCKGCFKAIVAGDQNVEYKGTVWHKDCFTCSNCKQV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.