missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6842-SP
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Western Blot.
Specifications
| FGFR2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGFR2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| 25 μg | |
| Primary | |
| Detects all isoforms of human FGF R2 in direct ELISAs and Western blots. Does not cross-react with any isoform of mouse FGF R2 or human FGF R1, FGF R3, or FGF R4. | |
| Human | |
| Purified |
| Western Blot | |
| 98742 | |
| Western Blot 1 ug/mL | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2263 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction