missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£181.00 - £1555.00
Specifications
| Antigen | FGFR2 |
|---|---|
| Clone | 98739 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.1-0.5 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753867
![]() |
R&D Systems
MAB6843-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702868
![]() |
R&D Systems
MAB6843-100 |
100 μg |
£392.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783358
![]() |
R&D Systems
MAB6843-500 |
500 μg |
£1555.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.Specifications
| FGFR2 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.1-0.5 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| FGFR2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 98739 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2263 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects all isoforms of human FGF R2 ( alpha and beta , IIIb and IIIc). In direct ELISAs, approximately 25-50% cross-reactivity with mouse FGF R2 and no cross-reactivity with any isoform of recombinant human (rh) FGF R1, rhFGF R3, or rhFGF R4 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title