missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGFR1a (IIIc) Fc Recombinant Protein, PeproTech®

Product Code. 17890523
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17890523

Brand: Gibco™ 16002100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGFR1a (IIIc) is a 65.2 kDa protein containing 586 amino acids. Under reducing conditions, FGFR1a migrates between 100-110 kDa on SDS-PAGE gel. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The FGF family plays a central role during prenatal development and postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. The FGF ligands bind to a family of type I transmembrane tyrosine kinase receptors, which leads to dimerization and activation by sequential autophosphorylation of specific tyrosine residues. Four genes encoding structurally related FGF receptors (FGFR-1 to -4) are known. Alternative splicing of the mRNAs generates numerous forms of FGFR-1 to -3. Alternate forms of FGF receptors can exhibit different specificities with respect to ligand binding. For example, the form designated as FGFR1a (IIc) interacts predominantly with FGF-acidic (FGF1) and FGF-basic (FGF2). A frequent splicing event involving FGFR-1 and -2 results in receptors containing all three Ig domains, referred to as the alpha isoform, or only IgII and IgIII, referred to as thebeta isoform. Only the alpha isoform has been identified for FGFR-3 and FGFR-4. Additional splicing events for FGFR-1 to -3, involving the C-terminal half of the IgIII domain encoded by two mutually exclusive alternative exons, generate FGF receptors with alternative IgIII domains (IIIb and IIIc).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11362
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2260
Molecular Weight (g/mol) 65.2 kDa
Name Human FGFR1a (IIIc) Fc
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name FGFR1a (IIIc)
Gene Symbol FGFR1
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is = 0.1 ng/ml, corresponding to a specific activity of = 1 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence RPSPTLPEQA QPWGAPVEVE SFLVHPGDLL QLRCRLRDDV QSINWLRDGV QLAESNRTRI TGEEVEVQDS VPADSGLYAC VTSSPSGSDT TYFSVNVSDA LPSSEDDDDD DDSSSEEKET DNTKPNPVAP YWTSPEKMEK KLHAVPAAKT VKFKCPSSGT PNPTLRWLKN SKEFKPDHRI GGYKVRYATW SIIMDSVVPS DKGNYTCIVE NEYGSINHTY QLDVVERSPH RPILQAGLPA NKTVALGSNV EFMCKVYSDP QPHIQWLKHI EVNGSKIGPD NLPYVQILKT AGVNTTDKEM EVLHLRNVSF EDAGEYTCLA GNSIGLSHHS AWLTVLEALE ERPAVMTSPL YLEGGPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.