missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR1 (IIIb) Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£167.00 - £1770.00
Specifications
| Antigen | FGFR1 |
|---|---|
| Clone | 133111 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.075-0.3 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726419
![]() |
R&D Systems
MAB765-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784498
![]() |
R&D Systems
MAB765-100 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753279
![]() |
R&D Systems
MAB765-500 |
500 μg |
£1770.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR1 Monoclonal specifically detects FGFR1 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| FGFR1 | |
| Western Blot 1 ug/mL, Neutralization 0.075-0.3 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGF R1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, Flt-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
| FGFR1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 133111 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2260 | |
| A mixture of purified isoforms of recombinant human FGF R1 extracellular domains | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects IIIb isoforms of human FGF R1. In direct ELISAs and Western blots, this antibody detected IIIb but not IIIc isoforms of human FGF R1, human FGF R2, and mouse FGF R2. In direct ELISA, approximately 20% cross-reactivity was observed with recombinant human (rh) FGF R3 (IIIb). No cross-reactivity was observed with rhFGF R4 or with IIIIc isoforms of rhFGF R1, rhFGF R2, rmFGF R2, or rhFGF R3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title