missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £1780.00
Specifications
| Antigen | FGFR1 |
|---|---|
| Clone | 133105 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737168
![]() |
R&D Systems
MAB658-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730928
![]() |
R&D Systems
MAB658-100 |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755589
![]() |
R&D Systems
MAB658-500 |
500 μg |
£1780.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR1 Monoclonal specifically detects FGFR1 in Human samples. It is validated for Western Blot.Specifications
| FGFR1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGF R1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, Flt-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
| FGFR1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 133105 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2260 | |
| Mouse myeloma cell line NS0-derived recombinant human FGF R1 alpha (IIIc) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FGF R1 in direct ELISAs and Western blots. Recognizes all isoforms of recombinant human (rh) FGF R1 and shows 10-25% cross-reactivity with various isoforms of rhFGF R2 and rhFGF R3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title