missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-BP Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB1593
This item is not returnable.
View return policy
Description
FGF-BP Monoclonal specifically detects FGF-BP in Human samples. It is validated for Western Blot.
Specifications
| FGF-BP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q14512 | |
| FGFBP1 | |
| E. coli-derived recombinant human FGF-BP Val35-Cys234 Accession # Q14512 | |
| 500 μg | |
| Primary | |
| Detects human FGF-BP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat FGF-BP is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 196519 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 17 kDa heparin-binding growth factor-binding protein, FGFBP, FGF-BP, FGFBP1, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9982 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction