missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems BAM233
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Specifications
| FGF basic/FGF2 | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2247 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 10043 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| FGF2 | |
| E. coli-derived recombinant human FGF basic | |
| 250 μg | |
| Primary | |
| Detects human FGF basic in ELISAs. In ELISAs, this antibody also recognizes bovine FGF basic but not bovine FGF acidic, recombinant human (rh) FGF acidic, rhFGF-4, rhFGF-6, or rhFGF-7. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction