missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGF-basic (FGF-2/bFGF) (147 aa) Recombinant Protein

Product Code. 11579141
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11579141 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11579141 Supplier Invitrogen™ Supplier No. RP8627

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Fibroblast Growth Factor basic (human) Recombinant Protein, RP-8627, from Invitrogen.

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization. Recombinant human FGF-b 147 (FGF-2) is a non-glycosylated protein, containing 147 amino acids, with a molecular weight of 16.5 kDa. Purity is typically greater than 97% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells and is typically less than 1ng/mL. The specific activity of this protein is 1x10^6 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -80°C and add 0.1% BSA for additional stability. Amino acid sequence: MPALPEDGGS GAFPPGHFKD PKRLYCKNGG FFLRIHPDGR VDGVREKSDP HIKLQLQAEE RGVVSIKGVC ANRYLAMKED GRLLASKCVT DECFFFERLE SNNYNTYRSR KYTSWYVALK RTGQYKLGSK TGPGQKAILF LPMSAKS.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09038
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 10mM sodium phosphate with 75mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 2247
Molecular Weight (g/mol) ∽16.5
Name Human FGF basic
pH Range 7.5
Quantity 50 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-FGF-b-147
Common Name Human FGF basic
Gene Symbol FGF2
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.