missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-8 Recombinant Protein
Click to view available options
Quantity:
25 μg
Unit Size:
Each
Description
Human FGF-8 Recombinant Protein for Bioactivity
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system. Plays a role in neurite outgrowth in hippocampal cells.
Specifications
Specifications
| Accession Number | P55075 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 2253 |
| Molecular Weight (g/mol) | 22.4 kDa |
| Name | Human FGF-8 |
| Purification Method | Purified |
| Quantity | 25 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction