missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-7 (KGF) Recombinant Protein

Product Code. 10631694
Click to view available options
Quantity:
10 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10631694

Brand: Gibco™ PHG0094

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute KGF in 1X PBS to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage, preferably desiccated. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21781
For Use With (Application) Bioactivity
Formulation PBS with 0.25M NaCl and no preservative; pH 7
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 18.9 kDa
Name Human FGF-7 (KGF)
pH Range 7
Purification Method Purified
Quantity 10 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Common Name KGF
Gene Symbol FGF7
Biological Activity ED50 = 0.1 - 15 ng/mL; determined by the dose-dependent proliferation of 4 MBr-5 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human FGF-7 (KGF) recombinant protein contains 163 amino acids
Content And Storage -20°C
Activity ED50 = 0.1 - 15 ng/mL; determined by the dose-dependent proliferation of 4MBr-5 cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype FGFs (Fibroblast Growth Factors)
Research Category Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis
Product Line Gibco
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.