missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB238-SP
This item is not returnable.
View return policy
Description
FGF-6 Monoclonal specifically detects FGF-6 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| FGF-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2251 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization, ELISA | |
| 27028 | |
| Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P10767 | |
| FGF6 | |
| E. coli-derived recombinant human FGF-6 Ala53-Ile208 Accession # P10767 | |
| 25 μg | |
| Primary | |
| Detects human FGF-6 in ELISAs. In direct ELISAs, approximately 75% cross-reactivity with recombinant human (rh) FGF-5 and less than 10% cross-reactivity with rhFGF-4 is observed. Does not neutralize the activity of FGF acidic, FGF basic, rhFGF-4, or rhFGF-5. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction