missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-6, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17806598
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
This item is not returnable. View return policy

Product Code. 17806598

Brand: Gibco™ AF100305UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGF-6 is an 18.7 kDa protein consisting of 168 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-6 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-6 is expressed in leukemia cell lines with platelet megakaryocytic differentiation potential. It signals through FGFR 1c, 2c, and 4.4
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10767
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 2251
Molecular Weight (g/mol) 18.7 kDa
Name Human FGF-6, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 5 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias FGF; FGF6; Fgf-6; Fibroblast growth factor; Fibroblast growth factor 6; HBGF-6; Heparin secretory-transforming protein 2; heparin-binding growth factor 6; HST2; HST-2; Hstf2; HSTF-2
Common Name FGF6
Gene Symbol FGF6
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence GTRANNTLLD SRGWGTLLSR SRAGLAGEIA GVNWESGYLV GIKRQRRLYC NVGIGFHLQV LPDGRISGTH EENPYSLLEI STVERGVVSL FGVRSALFVA MNSKGRLYAT PSFQEECKFR ETLLPNNYNA YESDLYQGTY IALSKYGRVK RGSKVSPIMT VTHFLPRI
Content And Storage -20°C
Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.