missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-22 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3867
This item is not returnable.
View return policy
Description
FGF-22 Monoclonal specifically detects FGF-22 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| FGF-22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF22, FGF-22, fibroblast growth factor 22 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27006 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization | |
| 435008 | |
| Western Blot 1 ug/mL, Neutralization 1.5-7.5 ug/mL | |
| Q9HCT0 | |
| FGF22 | |
| E. coli-derived recombinant human FGF-22 Thr23-Ser170 Accession # Q9HCT0 | |
| 500 μg | |
| Primary | |
| Detects human FGF-22 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FGF-basic, rhFGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -13, -16, -17, -18, -19, -20, -21, -23, recombinant mouse (rm) FGF-basic, rmFGF-8C, or -15 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction