missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-21 Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £500.00
Specifications
| Antigen | FGF-21 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795717
![]() |
R&D Systems
AF2539-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060792
|
R&D Systems
AF2539 |
100 μg |
£500.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-21 Polyclonal specifically detects FGF-21 in Human samples. It is validated for Western Blot, ELISA.Specifications
| FGF-21 | |
| Western Blot 0.1 ug/mL, ELISA | |
| Polyclonal | |
| Goat | |
| Human | |
| FGF21, FGF-21, fibroblast growth factor 21 | |
| FGF21 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Q9NSA1 | |
| 26291 | |
| E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human FGF-21 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse (rm) FGF-21 is observed and less than 2% cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, rmFGF-8b, rhFGF-9, rhFGF-10, rhFGF-11, rhFGF-12, rmFGF-15, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF-19, rhFGF-20, rhFGF-23, rhFGF acidic, and rhFGF basic is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title