missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-20 Recombinant Protein, PeproTech®
Description
100-41-1MG will be provided as 2 x 500 μg (100-41-500UG). Recombinant Human FGF-20 is a 23.2 kDa protein containing 209 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
Specifications
Specifications
| Accession Number | Q9NP95 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 26281 |
| Molecular Weight (g/mol) | 23.2 |
| Name | Human FGF-20 |
| Quantity | 2 x 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?