Learn More
Human FGF-18 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6671-100
 View more versions of this product
Product Code. 18714514
  • £460.00 / 100µg
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF18
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FGF18, FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
E. coli-derived human FGF-18 protein, Glu28-Ala207, Accession # O76093
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1
Immunohistochemistry
91811
Immunohistochemistry 5-15 μg/mL
O76093
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
8817
Human
Purified