missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-17 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF319-SP
This item is not returnable.
View return policy
Description
FGF-17 Polyclonal specifically detects FGF-17 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| FGF-17 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF-13, FGF17, FGF-17, fibroblast growth factor 17 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8822 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 6-24 ug/mL | |
| O60258 | |
| FGF17 | |
| E. coli-derived recombinant human FGF-17 Thr23-Thr216 Accession # O60258 | |
| 25 μg | |
| Primary | |
| Detects human FGF-17 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) FGF-8b and recombinant mouse FGF-8c is observed and less than 2% cross-reactivity with rhFGF acidic, rhFGF basic, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, rhFGF-10, and rhFGF-18 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction