missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-16 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
100 μg
2 x 250 μg
25 μg
250 μg
4 x 250 μg
5 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Pack of 2
Pack of 4
Description
100-29-1MG will be provided as 4 x 250 μg (100-29-250UG). 100-29-500UG will be provided as 2 x 250 μg (100-29-250UG). Recombinant Human FGF-16 is a 23.6 kDa protein consisting of 206 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.
Specifications
Specifications
| Accession Number | O43320 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 8823 |
| Molecular Weight (g/mol) | 23.6 kDa |
| Name | Human FGF-16 |
| Quantity | 2 x 250 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction