missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-16 Recombinant Protein, PeproTech®

Product Code. 17830673
Change view
Click to view available options
Quantity:
100 μg
2 x 250 μg
25 μg
250 μg
4 x 250 μg
5 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Pack of 2
Pack of 4
Product Code. Quantity unitSize
17830673 100 μg 100µg
17830883 4 x 250 μg 1mg
30088064 4 x 250 μg Pack of 4
17861753 250 μg 250µg
30087426 2 x 250 μg Pack of 2
17890943 25 μg 25µg
17870703 2 x 250 μg 500µg
17820953 5 μg 5µg
8 options
This item is not returnable. View return policy

Product Code. 17830673

Brand: Gibco™ 10029100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

100-29-1MG will be provided as 4 x 250 μg (100-29-250UG). 100-29-500UG will be provided as 2 x 250 μg (100-29-250UG). Recombinant Human FGF-16 is a 23.6 kDa protein consisting of 206 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43320
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8823
Molecular Weight (g/mol) 23.6 kDa
Name Human FGF-16
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4
Common Name FGF16
Gene Symbol FGF16
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 10 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.