missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-13 1B Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1909G-100UG
This item is not returnable.
View return policy
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human samples. It is validated for Flow Cytometry.
Specifications
| FGF-13 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FGF13, FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| E. coli-derived recombinant human FGF-13 isoform 1B Ala2-Thr192 Accession # NP_378668 | |
| RUO | |
| 2258 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 298923 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_378668 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FGF-13 1B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -16, -17, -19, -20, -21, rhFGF basic, recombinant mouse FGF-8b, or -8c is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction