missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FFAR1/GPR40 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £446.00
Specifications
| Antigen | FFAR1/GPR40 |
|---|---|
| Clone | 1048036 |
| Dilution | Immunohistochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791097
![]() |
R&D Systems
MAB11025-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792567
![]() |
R&D Systems
MAB11025-100 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FFAR1/GPR40 Monoclonal antibody specifically detects FFAR1/GPR40 in Human samples. It is validated for ImmunohistochemistrySpecifications
| FFAR1/GPR40 | |
| Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| FFA1, free fatty acid receptor 1, G protein-coupled receptor 40, GPCR40, GPR40, GPR40FFA1R, G-protein coupled receptor 40 | |
| Synthetic peptide within aminoacids 1-50 of human FFAR1/GPR40,, Accession # O14842 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1048036 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14842 | |
| 2864 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title