missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fetuin B Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB17251
This item is not returnable.
View return policy
Description
Fetuin B Monoclonal specifically detects Fetuin B in Human samples. It is validated for Western Blot, Simple Western, ELISA Capture (Matched Antibody Pair).
Specifications
| Fetuin B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 5 ug/mL, Simple Western 50 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9UGM5 | |
| FETUB | |
| Mouse myeloma cell line NS0-derived recombinant human Fetuin B Met19-Pro382 Accession # Q9UGM5 | |
| 500 μg | |
| Primary | |
| Detects human Fetuin B in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Fetuin A, rhCystatin A, B, C, rhMMP-2, 9, rhBMP-2, 4, 6, rhTGF-beta 1, or rhTGF-beta 2 is observed. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 212607 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26998 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction