missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Fetuin A/AHSG Recombinant Protein, PeproTech®

Product Code. 17801523
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17801523

Brand: Gibco™ 1401310UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Fetuin A is a heterodimeric protein containing a 282 amino acid A chain and a 27 amino acid B chain. The calculated molecular weight of Recombinant Human Fetuin A is 32.9 kDa. Due to glycosylation, recombinant Fetuin A migrates at an apparent molecular weight of 45-55 kDa by SDS-PAGE run under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02765
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 197
Molecular Weight (g/mol) 32.9 kDa
Name Human Fetuin A/AHSG
Quantity 10 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Common Name Fetuin A
Gene Symbol AHSG
Biological Activity Determined by its ability to inhibit Cathespin V cleavage of a fluorogenic peptide substrate Z-LR-AMC (RLUs). The expected IC50 is ≤ 100 nM.
Conjugate Unconjugated
Recombinant Recombinant
Sequence A-Chain: APHGPGLIYR QPNCDDPETE EAALVAIDYI NQNLPWGYKH TLNQIDEVKV WPQQPSGELF EIEIDTLETT CHVLDPTPVA RCSVRQLKEH AVEGDCDFQL LKLDGKFSVV YAKCDSSPDS AEDVRKVCQD CPLLAPLNDT RVVHAAKAAL AAFNAQNNGS NFQLEEISRA QLVPLPPSTY VEFTVSGTDC VAKEATEAAK CNLLAEKQYG FCKATLSEKL GGAEVAVTCT VFQTQPVTSQ PQPEGANEAV PTPVVDPDAP PSPPLGAPGL PPAGSPPDSH VL B-Chain: TVVQPSVG AAAGPVVPPC PGRIRHFKV
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.