missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fes Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF6460
This item is not returnable.
View return policy
Description
Fes Polyclonal specifically detects Fes in Human samples. It is validated for Western Blot.
Specifications
| Fes | |
| Polyclonal | |
| Unconjugated | |
| P07332 | |
| FES | |
| E. coli-derived recombinant human Fes aa 169-370 Accession # P07332 | |
| 100 μg | |
| Primary | |
| Detects human Fes in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| EC 2.7.10, EC 2.7.10.2, feline sarcoma oncogene, feline sarcoma virus, FPS, Proto-oncogene c-Fes, Proto-oncogene c-Fps, proto-oncogene tyrosine-protein kinase Fes/Fps, tyrosine-protein kinase Fes/Fps | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2242 | |
| Reconstitute at 0.2 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction