missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ferroportin/SLC40A1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9924-100
This item is not returnable.
View return policy
Description
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Ferroportin/SLC40A1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, SLC40A1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
| Synthetic peptide containing human Ferroportin/SLC40A1 Accession # Q9NP59 | |
| 100 μg | |
| 30061 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 1308C | |
| Flow Cytometry 0.25 μg/mL, Immunohistochemistry 1-25 μg/mL, CyTOF-ready | |
| Q9NP59 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Ferroportin/SLC40A1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction