missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fer Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7229
This item is not returnable.
View return policy
Description
Fer Monoclonal specifically detects Fer in Human samples. It is validated for Immunohistochemistry.
Specifications
| Fer | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P16591 | |
| FER | |
| E. coli-derived recombinant human Fer Asn93-Ala302 Accession # P16591 | |
| 100 μg | |
| Primary | |
| Detects human Fer in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 690318 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Phosphoprotein NCP94, Proto-oncogene c-Fer, TYK3, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2241 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction