missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRLB/FCRY Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4705N-100UG
This item is not returnable.
View return policy
Description
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FCRLB/FCRY | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6BAA4 | |
| FCRLB | |
| Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY Ala18-Ser426 Accession # Q6BAA4 | |
| RUO | |
| 127943 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 454217 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCRLB/FCRY in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction