missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL6/FcRH6 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6745R-100UG
This item is not returnable.
View return policy
Description
FCRL6/FcRH6 Monoclonal specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry.
Specifications
| FCRL6/FcRH6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6DN72 | |
| FCRL6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 Lys16-Leu312 Accession # Q6DN72 | |
| RUO | |
| 343413 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 672505 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Fc receptor-like 6, FcRH6, FLJ16056, IFGP6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCRL6/FcRH6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FCRL1, rhFCRL2, rhFCRL3, rhFCRL4, rhFCRL5, rhFcRN, or rhFCRLB/FCRY is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction