missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL6/FcRH6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6745G-100UG
This item is not returnable.
View return policy
Description
FCRL6/FcRH6 Monoclonal specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry.
Specifications
| FCRL6/FcRH6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Fc receptor-like 6, FcRH6, FLJ16056, IFGP6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human FCRL6/FcRH6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FCRL1, rhFCRL2, rhFCRL3, rhFCRL4, rhFCRL5, rhFcRN, or rhFCRLB/FCRY is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 672505 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q6DN72 | |
| FCRL6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6, Lys16-Leu312, Accession # Q6DN72 | |
| 100 μg | |
| 343413 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction