missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL5/FcRH5 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2087
This item is not returnable.
View return policy
Description
FCRL5/FcRH5/IRTA2 Polyclonal specifically detects FCRL5/FcRH5/IRTA2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| FCRL5/FcRH5/IRTA2 | |
| Polyclonal | |
| Unconjugated | |
| AAK93971 | |
| FCRL5 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL5/FcRH5 Gln16-Arg844 Accession # AAK93971 | |
| 100 μg | |
| Primary | |
| Detects human FCRL5/FcRH5 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IRTA3, rhIRTA4, and rhIRTA5 is observed and less than 5% cross-reactivity with rhIRTA1 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| BXMAS1, BXMAS1MGC119592, CD307, CD307e, CD307e antigen, DKFZp667E2019, DKFZp667F216, Fc receptor homolog 5, Fc receptor-like 5, Fc receptor-like protein 5, FcRH5, FCRH5MGC119593, FcRL5, FcR-like protein 5, FLJ00333, FLJ00397, IFGP5, Immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 (IRTA2), IRTA2, IRTA2MGC119590, PRO820 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 83416 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction