missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FCRL3/FcRH3 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Packungsgröße:
Each
Pack of 96
Beschreibung
Human FCRL3/FcRH3 quantitates human FCRL3/FcRH3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FCRL3/FcRH3.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Spezifikation
Spezifikation
| Accession Number | CD307c; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like protein 3; FCRH3; FCRL3; fcR-like protein 3; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; SH2 domain-containing phosphatase anchor protein 2; SPAP2 |
| Assay Range | 0.05 to 10 ng/mL |
| Assay Sensitivity | 0.05 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 115352 |
| Gene Symbol | FCRL3 |
| Interassay CV | <12% |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur