missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FCGR3B (O75015, Met 18 - Gly 193) Partial Recombinant Protein expressed in HEK293 cells

Product Code. 16251980
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 16251980

Brand: Abnova™ P3676.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

Specifications

Accession Number O75015
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 2215
Molecular Weight (g/mol) 50kDa
Name FCGR3B (Human) Recombinant Protein
Preparation Method Mammalian cell (HEK293) expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Storage Requirements Store at 4°C. For long term storage store at -20°C to -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CD16/CD16b/FCG3/FCGR3
Common Name FCGR3B
Gene Symbol FCGR3B
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (HEK293) expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.