missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc mu R/FAIM3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9494T-100UG
This item is not returnable.
View return policy
Description
FAIM3 Monoclonal specifically detects FAIM3 in Human samples. It is validated for Flow Cytometry.
Specifications
| FAIM3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FAIM3, Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, Toso, TOSOIgM Fc receptor | |
| Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession No. O60667 | |
| 100 μg | |
| 9214.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 992338 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| O60667 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Fc mu R/FAIM3 in direct ELISAs. Detects human, mouse, and rat Fc mu R/FAIM3 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction