missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIIA/B (CD16) Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1597
Additional Details : Weight : 0.00970kg
Description
Fc gamma RIII (CD16) Polyclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
Fc gamma RIII (CD16) | |
Polyclonal | |
Unconjugated | |
O75015 | |
FCGR3B | |
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIB (R&D Systems, Catalog # 1597-FC) Thr20-Gln208 Accession # O75015 | |
100 μg | |
Primary | |
Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human Fc gamma RIIA is observed and less than 2% cross-reactivity with recombinant mouse CD16 is observed. | |
Human | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction | |
CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20 | |
Goat | |
Affinity Purified | |
RUO | |
2214 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only