missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Fc gamma RIIA (CD32a) (167 Arg/His) His-tag Recombinant Protein

Description
Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | P12318 |
| For Use With (Application) | Control, Binding Assay, Immunohistochemistry, Surface Plasmon Resonance, Western Blot |
| Formulation | PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Gene ID (Entrez) | 2212 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | Human Fc gamma RIIA (CD32a) (167 Arg/His) His-tag |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | HEK293 cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?