missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RI/CD64 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB12571P
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry.
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2209 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 276426 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P12314.2 | |
| FCGR1A | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI Gln16-Pro288 Accession # P12314.2 | |
| 100 Tests | |
| Primary | |
| Detects human Fc gamma RI in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) Fc gamma RI, rmFc gamma RII, recombinant human (rh) Fc gamma RIIA, rhFc gamma RIIB, or rhFc gamma RIIIB. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction