missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human FBXW5 (aa 412-523) Control Fragment Recombinant Protein

Artikelnummer. 30195941
Klik for at se tilgængelige muligheder
Quantity:
100 μL
Pakningsstørrelse:
100µL
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30195941

Brand: Invitrogen™ RP102197

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61259 (PA5-61259. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains WD-40 domains, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene, however, they were found to be nonsense-mediated mRNA decay (NMD) candidates, hence not represented.
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Q969U6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54461
Name Human FBXW5 (aa 412-523) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI159739; F-box and WD repeat domain containing 5; F-box and WD-40 domain protein 5; F-box and WD-40 domain-containing protein 5; F-box protein Fbw5; F-box/WD repeat-containing protein 5; FBW5; Fbxw5; PP3971; WD repeat-containing F-box protein FBW5
Common Name FBXW5
Gene Symbol FBXW5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PDNRYLYVNSRAWPNGAVVADPMQPPPIAEEIDLLVFDLKTMREVRRALRAHRAYTPNDECFFIFLDVSRDFVASGAEDRHGYIWDRHYNICLARLRHEDVVNSVVFSPQEQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.